Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Fisherbrand™ Polyurethane Tractor Stool - Desk Height with Casters in Polyurethane
SCI_ED
Catalog No. S41249 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S41249 Blue
S41248 Black
15-000-743 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S41249
Add to Cart
Add to Cart
Fisherbrand™ Polyurethane Tractor Stools are height adjustable for 31 in. work surfaces and feature single-lever control mechanism and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant, easily cleaned, and features breathable holes for added comfort.
  • Adjustable from 16-21 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Product Type Tractor Stool
With Lumbar Support No
Weight (English) 24 lb.
Foot Type Black dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
With Armrest No
Load Bearing Capacity (English) 300 lb.
With Casters Yes
Adjustable (Height) 16 to 21 in.
Adjustable Yes
Color Blue
Base Material Nylon Reinforced Fiberglass
Load Bearing Capacity (Metric) 136 kg
With Backrest No
Footrest No
With Tilt No
Material Polyurethane
Seating Type Stool
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.